Assay Details
| Gene Symbol: LOC4347547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756907.2 | XP_015612393.1 | 11 - 12 | 1206 | 120 | |
| GenBank mRNA | AK066336.1 | - | 11 - 12 | 1214 | 120 | |
More Information
Set Membership: |
|

LOC4347547
Genomic Map