Assay Details
| Gene Symbol: LOC4347751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795679.2 | XP_015651165.1 | 1 - 2 | 456 | 75 | |
| GenBank mRNA | AK073737.1 | - | 1 - 2 | 430 | 75 | |
| AK105420.1 | - | 1 - 2 | 432 | 75 | ||
More Information
Set Membership: | 
		   
					
						
							 | 
				

LOC4347751
Genomic Map