Assay Details
| Gene Symbol: LOC4347834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756682.2 | XP_015612168.1 | 1 - 2 | 305 | 85 | |
| GenBank mRNA | AK102179.1 | - | 1 - 2 | 229 | 85 | |
More Information
Set Membership: |
|

LOC4347834
Genomic Map