Assay Details
| Gene Symbol: LOC9270313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756118.2 | XP_015611604.1 | 1 - 2 | 504 | 72 | |
| GenBank mRNA | AK318529.1 | - | 1 - 2 | 463 | 72 | |
More Information
Set Membership: |
|

LOC9270313
Genomic Map