Assay Details
| Gene Symbol: LOC4347947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759540.2 | XP_015615026.1 | 3 - 4 | 700 | 86 | |
| GenBank mRNA | AK106544.1 | - | 3 - 4 | 569 | 86 | |
More Information
Set Membership: |
|

LOC4347947
Genomic Map