Assay Details
| Gene Symbol: LOC4347999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759017.2 | XP_015614503.1 | 9 - 10 | 3085 | 127 | |
| GenBank mRNA | AK063350.1 | - | 4 - 5 | 437 | 127 | |
More Information
Set Membership: |
|

LOC4347999
Genomic Map