Assay Details
| Gene Symbol: LOC4347999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759017.2 | XP_015614503.1 | 11 - 12 | 3207 | 114 | |
| GenBank mRNA | AK063350.1 | - | 6 - 7 | 559 | 114 | |
More Information
Set Membership: |
|

LOC4347999
Genomic Map