Assay Details
| Gene Symbol: LOC4347999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759017.2 | XP_015614503.1 | 14 - 15 | 3436 | 134 | |
| GenBank mRNA | AK063350.1 | - | 9 - 10 | 788 | 134 | |
More Information
Set Membership: |
|

LOC4347999
Genomic Map