Assay Details
| Gene Symbol: LOC4348836 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759004.2 | XP_015614490.1 | 3 - 4 | 1920 | 151 | |
| GenBank mRNA | AK072090.1 | - | 3 - 4 | 1669 | 151 | |
| AK111526.1 | - | 3 - 4 | 1622 | 151 | ||
| AK111822.1 | - | 2 - 3 | 3400 | 151 | ||
More Information
Set Membership: |
|

LOC4348836
Genomic Map