Assay Details
| Gene Symbol: LOC4348984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758667.2 | XP_015614153.1 | 5 - 6 | 464 | 65 | |
| GenBank mRNA | AK240700.1 | - | 5 - 6 | 445 | 65 | |
| Gene Symbol: LOC4335698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK240828.1 | - | 1960 | 65 | ||
More Information
Set Membership: |
|

LOC4348984
Genomic Map