Assay Details
| Gene Symbol: LOC4348984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758667.2 | XP_015614153.1 | 6 - 7 | 537 | 69 | |
| GenBank mRNA | AK240700.1 | - | 6 - 7 | 518 | 69 | |
| Gene Symbol: LOC4335698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK240828.1 | - | 1887 | 69 | ||
More Information
Set Membership: |
|

LOC4348984
Genomic Map