Assay Details
| Gene Symbol: LOC4349237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759313.2 | XP_015614799.1 | 3 - 4 | 772 | 120 | |
| GenBank mRNA | AK070135.1 | - | 3 - 4 | 538 | 120 | |
| AY345224.1 | - | 3 - 4 | 542 | 120 | ||
| AY551919.1 | - | 2 - 3 | 375 | 120 | ||
More Information
Set Membership: |
|

LOC4349237
Genomic Map