Assay Details
| Gene Symbol: LOC4349273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759406.2 | XP_015614892.1 | 1 - 2 | 229 | 147 | |
| GenBank mRNA | AK073941.1 | - | 1 - 2 | 199 | 147 | |
More Information
Set Membership: |
|

LOC4349273
Genomic Map