Assay Details
| Gene Symbol: LOC4349273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759406.2 | XP_015614892.1 | 2 - 3 | 478 | 73 | |
| GenBank mRNA | AK060182.1 | - | 1 - 2 | 71 | 73 | |
| AK073941.1 | - | 2 - 3 | 448 | 73 | ||
More Information
Set Membership: |
|

LOC4349273
Genomic Map