Assay Details
| Gene Symbol: LOC4349273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759406.2 | XP_015614892.1 | 3 - 4 | 618 | 153 | |
| GenBank mRNA | AK060182.1 | - | 2 - 3 | 211 | 153 | |
| AK073941.1 | - | 3 - 4 | 588 | 153 | ||
More Information
Set Membership: |
|

LOC4349273
Genomic Map