Assay Details
| Gene Symbol: LOC4349276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758722.2 | XP_015614208.1 | 2 - 3 | 905 | 142 | |
| GenBank mRNA | AK069472.1 | - | 1 - 2 | 965 | 142 | |
More Information
Set Membership: |
|

LOC4349276
Genomic Map