Assay Details
| Gene Symbol: LOC4349276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758722.2 | XP_015614208.1 | 3 - 4 | 1196 | 109 | |
| GenBank mRNA | AK069472.1 | - | 2 - 3 | 1256 | 109 | |
More Information
Set Membership: |
|

LOC4349276
Genomic Map