Assay Details
| Gene Symbol: LOC4349776 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761750.2 | XP_015617236.1 | 4 - 5 | 3810 | 85 | |
| GenBank mRNA | AK066721.1 | - | 2 - 3 | 749 | 85 | |
More Information
Set Membership: |
|

LOC4349776
Genomic Map