Assay Details
| Gene Symbol: LOC4349776 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761750.2 | XP_015617236.1 | 7 - 8 | 4420 | 95 | |
| GenBank mRNA | AK066721.1 | - | 5 - 6 | 1359 | 95 | |
More Information
Set Membership: |
|

LOC4349776
Genomic Map