Assay Details
| Gene Symbol: LOC4351629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762718.2 | XP_015618204.1 | 8 - 9 | 1089 | 70 | |
| XM_015762719.2 | XP_015618205.1 | 8 - 9 | 1089 | 70 | ||
| GenBank mRNA | AK107402.1 | - | 8 - 9 | 1041 | 70 | |
More Information
Set Membership: |
|

LOC4351629
Genomic Map