Assay Details
| Gene Symbol: LOC4351629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762718.2 | XP_015618204.1 | 9 - 10 | 1185 | 135 | |
| GenBank mRNA | AK107402.1 | - | 9 - 10 | 1137 | 135 | |
More Information
Set Membership: |
|

LOC4351629
Genomic Map