Assay Details
| Gene Symbol: LOC4351679 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764960.2 | XP_015620446.1 | 1 - 2 | 2906 | 75 | |
| GenBank mRNA | AK067635.1 | - | 1 - 2 | 2793 | 75 | |
More Information
Set Membership: |
|

LOC4351679
Genomic Map