Assay Details
| Gene Symbol: LOC4351709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764965.2 | XP_015620451.1 | 10 - 11 | 1388 | 95 | |
| GenBank mRNA | AK100343.1 | - | 10 - 11 | 1389 | 95 | |
More Information
Set Membership: |
|

LOC4351709
Genomic Map