Assay Details
| Gene Symbol: LOC4351709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764965.2 | XP_015620451.1 | 13 - 14 | 1775 | 107 | |
| GenBank mRNA | AK100343.1 | - | 13 - 14 | 1776 | 107 | |
More Information
Set Membership: |
|

LOC4351709
Genomic Map