Assay Details
| Gene Symbol: LOC4351709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764965.2 | XP_015620451.1 | 1 - 2 | 286 | 107 | |
| GenBank mRNA | AK100343.1 | - | 1 - 2 | 287 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015764965.2 transcripts.
Set Membership: |
|

LOC4351709
Genomic Map