Assay Details
| Gene Symbol: LOC4351752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762954.2 | XP_015618440.1 | 2 - 3 | 278 | 103 | |
| GenBank mRNA | AY177700.1 | - | 2 - 3 | 264 | 103 | |
More Information
Set Membership: |
|

LOC4351752
Genomic Map