Assay Details
| Gene Symbol: LOC4351828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764598.2 | XP_015620084.1 | 7 - 8 | 1570 | 99 | |
| XM_015764599.2 | XP_015620085.1 | 7 - 8 | 1550 | 99 | ||
| GenBank mRNA | AK066153.1 | - | 7 - 8 | 1549 | 99 | |
More Information
Set Membership: |
|

LOC4351828
Genomic Map