Assay Details
| Gene Symbol: LOC4351828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764598.2 | XP_015620084.1 | 9 - 10 | 1731 | 99 | |
| XM_015764599.2 | XP_015620085.1 | 9 - 10 | 1711 | 99 | ||
| GenBank mRNA | AK066153.1 | - | 9 - 10 | 1710 | 99 | |
More Information
Set Membership: |
|

LOC4351828
Genomic Map