Assay Details
| Gene Symbol: LOC4351908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762694.2 | XP_015618180.1 | 12 - 13 | 1529 | 61 | |
| XM_015762695.2 | XP_015618181.1 | 12 - 13 | 1524 | 61 | ||
| GenBank mRNA | AK062220.1 | - | 1 - 2 | 123 | 61 | |
| AK064874.1 | - | 12 - 13 | 1447 | 61 | ||
| AK072798.1 | - | 9 - 10 | 975 | 61 | ||
More Information
Set Membership: |
|

LOC4351908
Genomic Map