Assay Details
| Gene Symbol: LOC4351908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762694.2 | XP_015618180.1 | 2 - 3 | 445 | 58 | |
| XM_015762695.2 | XP_015618181.1 | 2 - 3 | 443 | 58 | ||
| GenBank mRNA | AK064874.1 | - | 2 - 3 | 366 | 58 | |
More Information
Set Membership: |
|

LOC4351908
Genomic Map