Assay Details
| Gene Symbol: LOC4351908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762694.2 | XP_015618180.1 | 7 - 8 | 889 | 71 | |
| XM_015762695.2 | XP_015618181.1 | 7 - 8 | 884 | 71 | ||
| GenBank mRNA | AK072798.1 | - | 4 - 5 | 335 | 71 | |
More Information
Set Membership: |
|

LOC4351908
Genomic Map