Assay Details
| Gene Symbol: LOC4352624 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763692.2 | XP_015619178.1 | 5 - 6 | 867 | 62 | |
| GenBank mRNA | AK105512.1 | - | 5 - 6 | 835 | 62 | |
More Information
Set Membership: |
|

LOC4352624
Genomic Map