Assay Details
| Gene Symbol: LOC4352674 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763074.2 | XP_015618560.1 | 17 - 18 | 2449 | 84 | |
| XM_015763076.2 | XP_015618562.1 | 17 - 18 | 2449 | 84 | ||
More Information
Set Membership: |
|

LOC4352674
Genomic Map