Assay Details
| Gene Symbol: LOC4352674 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763074.2 | XP_015618560.1 | 23 - 24 | 4666 | 136 | |
| XM_015763076.2 | XP_015618562.1 | 23 - 24 | 4666 | 136 | ||
More Information
Set Membership: |
|

LOC4352674
Genomic Map