Assay Details
| Gene Symbol: LOC4352674 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763076.2 | XP_015618562.1 | 31 - 32 | 7851 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4352674| Gene Symbol: LOC4352674 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763076.2 | XP_015618562.1 | 31 - 32 | 7851 | 96 | |
Set Membership: |
|