Assay Details
Gene Symbol: LOC4352741 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765102.2 | XP_015620588.1 | 10 - 11 | 1491 | 84 | |
GenBank mRNA | AK061906.1 | - | 7 - 8 | 745 | 84 | |
AK065165.1 | - | 9 - 10 | 1454 | 84 | ||
AK121511.1 | - | 10 - 11 | 1453 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
