Assay Details
Gene Symbol: LOC4352741 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765102.2 | XP_015620588.1 | 12 - 13 | 1888 | 144 | |
GenBank mRNA | AF251074.1 | - | 1 - 2 | 142 | 144 | |
AK061906.1 | - | 9 - 10 | 1142 | 144 | ||
AK121511.1 | - | 12 - 13 | 1850 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
