Assay Details
Gene Symbol: LOC4352741 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765102.2 | XP_015620588.1 | 15 - 16 | 2343 | 82 | |
GenBank mRNA | AF251074.1 | - | 4 - 5 | 597 | 82 | |
AK061906.1 | - | 12 - 13 | 1597 | 82 | ||
AK065165.1 | - | 13 - 14 | 2387 | 82 | ||
AK121511.1 | - | 15 - 16 | 2305 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
