Assay Details
Gene Symbol: LOC4352741 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765102.2 | XP_015620588.1 | 4 - 5 | 886 | 143 | |
GenBank mRNA | AK061906.1 | - | 1 - 2 | 140 | 143 | |
AK065165.1 | - | 4 - 5 | 772 | 143 | ||
AK121511.1 | - | 4 - 5 | 848 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
