Assay Details
Gene Symbol: LOC4352741 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765102.2 | XP_015620588.1 | 7 - 8 | 1148 | 87 | |
GenBank mRNA | AK061906.1 | - | 4 - 5 | 402 | 87 | |
AK065165.1 | - | 6 - 7 | 1111 | 87 | ||
AK121511.1 | - | 7 - 8 | 1110 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
