Assay Details
Gene Symbol: LOC4352741 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765102.2 | XP_015620588.1 | 8 - 9 | 1252 | 67 | |
GenBank mRNA | AK061906.1 | - | 5 - 6 | 506 | 67 | |
AK065165.1 | - | 7 - 8 | 1215 | 67 | ||
AK121511.1 | - | 8 - 9 | 1214 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
