Assay Details
| Gene Symbol: LOC4350201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762236.2 | XP_015617722.1 | 5 - 6 | 930 | 113 | |
| GenBank mRNA | AK107293.1 | - | 4 - 5 | 315 | 113 | |
More Information
Set Membership: |
|

LOC4350201
Genomic Map