Assay Details
| Gene Symbol: LOC4350201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762236.2 | XP_015617722.1 | 7 - 8 | 1079 | 86 | |
| GenBank mRNA | AK107293.1 | - | 6 - 7 | 464 | 86 | |
More Information
Set Membership: |
|

LOC4350201
Genomic Map