Assay Details
| Gene Symbol: LOC4350241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759709.2 | XP_015615195.1 | 3 - 4 | 1225 | 99 | |
| GenBank mRNA | AK071935.1 | - | 3 - 4 | 1236 | 99 | |
| AK103706.1 | - | 3 - 4 | 1234 | 99 | ||
More Information
Set Membership: |
|

LOC4350241
Genomic Map