Assay Details
| Gene Symbol: LOC4350241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759709.2 | XP_015615195.1 | 4 - 5 | 1327 | 70 | |
| GenBank mRNA | AK071935.1 | - | 4 - 5 | 1338 | 70 | |
| AK103706.1 | - | 4 - 5 | 1336 | 70 | ||
More Information
Set Membership: |
|

LOC4350241
Genomic Map