Assay Details
| Gene Symbol: LOC4350244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759645.1 | XP_015615131.1 | 1 - 2 | 356 | 57 | |
| XM_015759646.2 | XP_015615132.1 | 1 - 2 | 359 | 57 | ||
More Information
Set Membership: |
|

LOC4350244
Genomic Map