Assay Details
| Gene Symbol: LOC4350258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762437.2 | XP_015617923.1 | 9 - 10 | 1778 | 110 | |
| GenBank mRNA | AK069552.1 | - | 9 - 10 | 1771 | 110 | |
| AK100126.1 | - | 9 - 10 | 1779 | 110 | ||
More Information
Set Membership: |
|

LOC4350258
Genomic Map