Assay Details
| Gene Symbol: LOC4350275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762497.2 | XP_015617983.1 | 4 - 5 | 625 | 73 | |
| GenBank mRNA | AK103343.1 | - | 4 - 5 | 562 | 73 | |
More Information
Set Membership: |
|

LOC4350275
Genomic Map