Assay Details
| Gene Symbol: LOC4350402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761860.2 | XP_015617346.1 | 10 - 11 | 1292 | 137 | |
| GenBank mRNA | AK067867.1 | - | 10 - 11 | 1766 | 137 | |
| AK100614.1 | - | 9 - 10 | 983 | 137 | ||
More Information
Set Membership: |
|

LOC4350402
Genomic Map