Assay Details
| Gene Symbol: LOC4350402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761860.2 | XP_015617346.1 | 11 - 12 | 1422 | 70 | |
| GenBank mRNA | AK067867.1 | - | 11 - 12 | 1896 | 70 | |
| AK100614.1 | - | 10 - 11 | 1113 | 70 | ||
More Information
Set Membership: |
|

LOC4350402
Genomic Map